Assay Details
| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002901.3 | NP_001002901.1 | 6 - 7 | 812 | 67 | |
| NM_001320241.1 | NP_001307170.1 | 5 - 6 | 626 | 67 | ||
| GenBank mRNA | AK290733.1 | - | 5 - 6 | 626 | 67 | |
| AY670683.1 | - | 4 - 5 | 593 | 67 | ||
| AY786314.1 | - | 4 - 5 | 578 | 67 | ||
| BC038564.2 | - | 1 - 2 | 178 | 67 | ||
| DQ021957.1 | - | 6 - 7 | 812 | 67 | ||
More Information
Set Membership: |
|

FCRLB
Genomic Map