Assay Details
| Gene Symbol: CUL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198777.1 | NP_001185706.1 | 17 - 18 | 1913 | 93 | |
| NM_001198778.1 | NP_001185707.1 | 17 - 18 | 1839 | 93 | ||
| NM_001198779.1 | NP_001185708.1 | 17 - 18 | 1952 | 93 | ||
| NM_001324375.1 | NP_001311304.1 | 15 - 16 | 1716 | 93 | ||
| NM_001324376.1 | NP_001311305.1 | 18 - 19 | 2073 | 93 | ||
| NM_003591.3 | NP_003582.2 | 17 - 18 | 1905 | 93 | ||
| XM_011519743.1 | XP_011518045.1 | 19 - 20 | 2067 | 93 | ||
| XM_011519744.1 | XP_011518046.1 | 18 - 19 | 1949 | 93 | ||
| XM_011519745.1 | XP_011518047.1 | 18 - 19 | 2001 | 93 | ||
| XM_011519747.1 | XP_011518049.1 | 18 - 19 | 1986 | 93 | ||
| GenBank mRNA | AF126404.1 | - | 16 - 17 | 1678 | 93 | |
| AK095217.1 | - | 17 - 18 | 1913 | 93 | ||
| AK294080.1 | - | 15 - 16 | 1664 | 93 | ||
| AK300491.1 | - | 17 - 18 | 1797 | 93 | ||
| AK310225.1 | - | 18 - 19 | 1984 | 93 | ||
| AL832733.1 | - | 17 - 18 | 1889 | 93 | ||
| BC009591.1 | - | 17 - 18 | 1888 | 4104375 | 93 | |
| BC110901.1 | - | 17 - 18 | 1893 | 6161736 | 93 | |
| U58088.1 | - | 14 - 15 | 1398 | 93 | ||
| U83410.1 | - | 17 - 18 | 1824 | 93 | ||
More Information
Set Membership: |
|

CUL2
Genomic Map