Assay Details
| Gene Symbol: CUL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198777.1 | NP_001185706.1 | 18 - 19 | 2118 | 187 | |
| NM_001198778.1 | NP_001185707.1 | 18 - 19 | 2044 | 187 | ||
| NM_001198779.1 | NP_001185708.1 | 18 - 19 | 2157 | 187 | ||
| NM_001324375.1 | NP_001311304.1 | 16 - 17 | 1921 | 187 | ||
| NM_001324376.1 | NP_001311305.1 | 19 - 20 | 2278 | 187 | ||
| NM_003591.3 | NP_003582.2 | 18 - 19 | 2110 | 187 | ||
| XM_011519743.1 | XP_011518045.1 | 20 - 21 | 2272 | 187 | ||
| XM_011519744.1 | XP_011518046.1 | 19 - 20 | 2154 | 187 | ||
| XM_011519745.1 | XP_011518047.1 | 19 - 20 | 2206 | 187 | ||
| XM_011519747.1 | XP_011518049.1 | 19 - 20 | 2191 | 187 | ||
| GenBank mRNA | AF126404.1 | - | 17 - 18 | 1883 | 187 | |
| AK095217.1 | - | 18 - 19 | 2118 | 187 | ||
| AK300491.1 | - | 18 - 19 | 2002 | 187 | ||
| AK310225.1 | - | 19 - 20 | 2189 | 187 | ||
| AL832733.1 | - | 18 - 19 | 2094 | 187 | ||
| BC009591.1 | - | 18 - 19 | 2093 | 4104375 | 187 | |
| BC110901.1 | - | 18 - 19 | 2098 | 6161736 | 187 | |
| U58088.1 | - | 15 - 16 | 1603 | 187 | ||
| U83410.1 | - | 18 - 19 | 2029 | 187 | ||
More Information
Set Membership: |
|

CUL2
Genomic Map