Assay Details
| Gene Symbol: CUL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198777.1 | NP_001185706.1 | 2 - 3 | 347 | 66 | |
| NM_001198778.1 | NP_001185707.1 | 2 - 3 | 273 | 66 | ||
| NM_001198779.1 | NP_001185708.1 | 2 - 3 | 386 | 66 | ||
| NM_001324375.1 | NP_001311304.1 | 2 - 3 | 339 | 66 | ||
| NM_001324376.1 | NP_001311305.1 | 2 - 3 | 386 | 66 | ||
| NM_003591.3 | NP_003582.2 | 2 - 3 | 339 | 66 | ||
| XM_011519743.1 | XP_011518045.1 | 4 - 5 | 501 | 66 | ||
| XM_011519744.1 | XP_011518046.1 | 3 - 4 | 383 | 66 | ||
| XM_011519745.1 | XP_011518047.1 | 3 - 4 | 435 | 66 | ||
| XM_011519747.1 | XP_011518049.1 | 3 - 4 | 420 | 66 | ||
| GenBank mRNA | AF126404.1 | - | 1 - 2 | 112 | 66 | |
| AK095217.1 | - | 2 - 3 | 347 | 66 | ||
| AK294080.1 | - | 2 - 3 | 287 | 66 | ||
| AK300491.1 | - | 2 - 3 | 231 | 66 | ||
| AK310225.1 | - | 2 - 3 | 297 | 66 | ||
| AL832733.1 | - | 2 - 3 | 323 | 66 | ||
| BC009591.1 | - | 2 - 3 | 322 | 4104375 | 66 | |
| BC110901.1 | - | 2 - 3 | 327 | 6161736 | 66 | |
| DC401811.1 | - | 2 - 3 | 274 | 66 | ||
| DC401936.1 | - | 2 - 3 | 339 | 66 | ||
| U83410.1 | - | 2 - 3 | 258 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001324376.1 transcripts.
Set Membership: |
|

CUL2
Genomic Map