Assay Details
| Gene Symbol: CUL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324375.1 | NP_001311304.1 | 4 - 5 | 543 | 77 | |
| GenBank mRNA | AK294080.1 | - | 4 - 5 | 491 | 77 | |
More Information
Set Membership: |
|

CUL2
Genomic Map