Assay Details
| Gene Symbol: CUL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198777.1 | NP_001185706.1 | 7 - 8 | 840 | 99 | |
| NM_001198778.1 | NP_001185707.1 | 7 - 8 | 766 | 99 | ||
| NM_001198779.1 | NP_001185708.1 | 7 - 8 | 879 | 99 | ||
| NM_001324375.1 | NP_001311304.1 | 5 - 6 | 643 | 99 | ||
| NM_001324376.1 | NP_001311305.1 | 8 - 9 | 1000 | 99 | ||
| NM_003591.3 | NP_003582.2 | 7 - 8 | 832 | 99 | ||
| XM_011519743.1 | XP_011518045.1 | 9 - 10 | 994 | 99 | ||
| XM_011519744.1 | XP_011518046.1 | 8 - 9 | 876 | 99 | ||
| XM_011519745.1 | XP_011518047.1 | 8 - 9 | 928 | 99 | ||
| XM_011519747.1 | XP_011518049.1 | 8 - 9 | 913 | 99 | ||
| GenBank mRNA | AF126404.1 | - | 6 - 7 | 605 | 99 | |
| AK095217.1 | - | 7 - 8 | 840 | 99 | ||
| AK294080.1 | - | 5 - 6 | 591 | 99 | ||
| AK300491.1 | - | 7 - 8 | 724 | 99 | ||
| AK310225.1 | - | 8 - 9 | 911 | 99 | ||
| BC009591.1 | - | 7 - 8 | 815 | 4104375 | 99 | |
| BC110901.1 | - | 7 - 8 | 820 | 6161736 | 99 | |
| U58088.1 | - | 4 - 5 | 325 | 99 | ||
| U83410.1 | - | 7 - 8 | 751 | 99 | ||
More Information
Set Membership: |
|

CUL2
Genomic Map