Assay Details
| Gene Symbol: CUL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198779.1 | NP_001185708.1 | 1 - 2 | 253 | 67 | |
| NM_001324376.1 | NP_001311305.1 | 1 - 2 | 253 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001198779.1, NM_001324376.1 transcripts.
Set Membership: |
|

CUL2
Genomic Map