Assay Details
| Gene Symbol: CUL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198777.1 | NP_001185706.1 | 5 - 6 | 659 | 101 | |
| NM_001198778.1 | NP_001185707.1 | 5 - 6 | 585 | 101 | ||
| NM_001198779.1 | NP_001185708.1 | 5 - 6 | 698 | 101 | ||
| NM_001324376.1 | NP_001311305.1 | 6 - 7 | 819 | 101 | ||
| NM_003591.3 | NP_003582.2 | 5 - 6 | 651 | 101 | ||
| XM_011519743.1 | XP_011518045.1 | 7 - 8 | 813 | 101 | ||
| XM_011519744.1 | XP_011518046.1 | 6 - 7 | 695 | 101 | ||
| XM_011519745.1 | XP_011518047.1 | 6 - 7 | 747 | 101 | ||
| XM_011519747.1 | XP_011518049.1 | 6 - 7 | 732 | 101 | ||
| GenBank mRNA | AF126404.1 | - | 4 - 5 | 424 | 101 | |
| AK095217.1 | - | 5 - 6 | 659 | 101 | ||
| AK300491.1 | - | 5 - 6 | 543 | 101 | ||
| AK310225.1 | - | 6 - 7 | 730 | 101 | ||
| AL832733.1 | - | 5 - 6 | 635 | 101 | ||
| BC009591.1 | - | 5 - 6 | 634 | 4104375 | 101 | |
| BC110901.1 | - | 5 - 6 | 639 | 6161736 | 101 | |
| U58088.1 | - | 2 - 3 | 144 | 101 | ||
| U83410.1 | - | 5 - 6 | 570 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001324376.1 transcripts.
Set Membership: |
|

CUL2
Genomic Map