Assay Details
| Gene Symbol: ZBED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011514283.2 | XP_011512585.1 | 3 - 4 | 475 | 75 | |
| XM_011514284.2 | XP_011512586.1 | 3 - 4 | 431 | 75 | ||
| XM_011514285.2 | XP_011512587.1 | 3 - 4 | 397 | 75 | ||
| XM_011514286.2 | XP_011512588.1 | 3 - 4 | 408 | 75 | ||
| XM_017010231.1 | XP_016865720.1 | 1 - 2 | 468 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011514283.2, XM_011514284.2, XM_011514285.2, XM_011514286.2, XM_017010231.1 transcripts.
Set Membership: |
|

ZBED9
Genomic Map