Assay Details
| Gene Symbol: FIGNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017012501.1 | XP_016867990.1 | 3 - 4 | 1720 | 93 | |
| XM_017012502.1 | XP_016867991.1 | 4 - 5 | 892 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017012501.1, XM_017012502.1 transcripts.
Set Membership: |
|

FIGNL1
Genomic Map