Assay Details
| Gene Symbol: LRRC4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080457.1 | NP_001073926.1 | 2 - 3 | 432 | 93 | |
| XM_005259429.4 | XP_005259486.1 | 2 - 3 | 611 | 93 | ||
| XM_006723506.3 | XP_006723569.1 | 3 - 4 | 713 | 93 | ||
| XM_011527520.2 | XP_011525822.1 | 2 - 3 | 1859 | 93 | ||
| GenBank mRNA | BC032460.1 | - | 1 - 2 | 307 | 93 | |
| BC131897.1 | - | 2 - 3 | 410 | 93 | ||
More Information
Set Membership: |
|

LRRC4B
Genomic Map