Assay Details
| Gene Symbol: BAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172415.1 | NP_001165886.1 | 5 - 6 | 966 | 132 | |
| NM_004323.5 | NP_004314.5 | 5 - 6 | 966 | 132 | ||
| GenBank mRNA | AF022224.1 | - | 5 - 6 | 924 | 132 | |
| AK093238.1 | - | 2 - 3 | 1625 | 132 | ||
| AK096038.1 | - | 6 - 7 | 729 | 132 | ||
| AK222749.1 | - | 5 - 6 | 913 | 132 | ||
| AK316577.1 | - | 5 - 6 | 892 | 132 | ||
| BC001936.1 | - | 5 - 6 | 927 | 2823774 | 132 | |
| BC014774.1 | - | 5 - 6 | 776 | 132 | ||
| KU177932.1 | - | 5 - 6 | 666 | 132 | ||
| KU177933.1 | - | 6 - 7 | 808 | 132 | ||
| Z35491.1 | - | 5 - 6 | 944 | 132 | ||
More Information
Set Membership: |
|

BAG1
Genomic Map