Assay Details
| Gene Symbol: SPINK14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001325.2 | NP_001001325.1 | 4 - 5 | 342 | 92 | |
| XM_017009466.1 | XP_016864955.1 | 4 - 5 | 415 | 92 | ||
| XM_017009468.1 | XP_016864957.1 | 4 - 5 | 829 | 92 | ||
| XM_017009469.1 | XP_016864958.1 | 3 - 4 | 536 | 92 | ||
| GenBank mRNA | JF509943.1 | - | 4 - 5 | 342 | 92 | |
| JF509944.1 | - | 2 - 3 | 254 | 92 | ||
More Information
Set Membership: |
|

SPINK14
Genomic Map