Assay Details
| Gene Symbol: MRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002438.3 | NP_002429.1 | ||||
| GenBank mRNA | AK308167.1 | - | ||||
| BC146838.1 | - | 9056765 | ||||
| DQ663787.1 | - | |||||
| J05550.1 | - | |||||
| X55635.1 | - | |||||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002438.3 | NP_002429.1 | ||||
| GenBank mRNA | AK308167.1 | - | ||||
| BC146838.1 | - | 9056765 | ||||
| DQ663787.1 | - | |||||
| J05550.1 | - | |||||
| X55635.1 | - | |||||