Assay Details
| Gene Symbol: MRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002438.3 | NP_002429.1 | 25 - 26 | 3769 | 99 | |
| GenBank mRNA | AK297044.1 | - | 9 - 10 | 1229 | 99 | |
| BC146838.1 | - | 25 - 26 | 3692 | 9056765 | 99 | |
| DQ663787.1 | - | 25 - 26 | 3646 | 99 | ||
| J05550.1 | - | 25 - 26 | 3749 | 99 | ||
| X55635.1 | - | 25 - 26 | 3722 | 99 | ||
More Information
Set Membership: |
|

MRC1
Genomic Map