Assay Details
| Gene Symbol: GPATCH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015590.3 | NP_056405.2 | 2 - 3 | 138 | 115 | |
| NM_182679.2 | NP_872620.1 | 2 - 3 | 212 | 115 | ||
| XM_005245287.3 | XP_005245344.1 | 2 - 3 | 221 | 115 | ||
| GenBank mRNA | AK000256.1 | - | 2 - 3 | 205 | 115 | |
| AK293026.1 | - | 2 - 3 | 206 | 115 | ||
| AK301232.1 | - | 2 - 3 | 204 | 115 | ||
| BC032436.1 | - | 2 - 3 | 191 | 115 | ||
| BC040147.1 | - | 2 - 3 | 195 | 115 | ||
| CB109133.1 | - | 2 - 3 | 121 | 115 | ||
| DA087471.1 | - | 2 - 3 | 212 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC040147.1 transcripts.
Set Membership: |
|

GPATCH4
Genomic Map