Assay Details
| Gene Symbol: RAB39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017516.2 | NP_059986.1 | 1 - 2 | 316 | 91 | |
| GenBank mRNA | AK292969.1 | - | 1 - 2 | 288 | 91 | |
| BC028064.1 | - | 1 - 2 | 240 | 5197104 | 91 | |
| HY107079.1 | - | 1 - 2 | 316 | 92 | ||
| X99962.1 | - | 1 - 2 | 222 | 91 | ||
More Information
Set Membership: |
|

RAB39A
Genomic Map