Assay Details
| Gene Symbol: SMIM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287445.1 | NP_001274374.1 | 1 - 2 | 46 | 70 | |
| NM_020425.5 | NP_065158.3 | 1 - 2 | 46 | 70 | ||
| GenBank mRNA | BG534658.1 | - | 1 - 2 | 47 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001287445.1, NM_020425.5 transcripts.
Set Membership: |
|

SMIM8
Genomic Map