Assay Details
| Gene Symbol: OVCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183378.2 | NP_899234.2 | 19 - 20 | 2282 | 97 | |
| XM_011520638.2 | XP_011518940.1 | 19 - 20 | 4394 | 97 | ||
| XM_011520639.2 | XP_011518941.1 | 19 - 20 | 4394 | 97 | ||
| XM_017019256.1 | XP_016874745.1 | 19 - 20 | 4394 | 97 | ||
| XM_017019257.1 | XP_016874746.1 | 19 - 20 | 4394 | 97 | ||
More Information
Set Membership: |
|

OVCH1
Genomic Map