Assay Details
| Gene Symbol: ZNF662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207404.3 | NP_997287.2 | 3 - 4 | 495 | 87 | |
| GenBank mRNA | AK127779.1 | - | 3 - 4 | 495 | 87 | |
More Information
Set Membership: |
|

ZNF662
Genomic Map