Assay Details
| Gene Symbol: SNX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144972.1 | NP_001138444.1 | 2 - 3 | 299 | 89 | |
| NM_153337.2 | NP_699168.1 | 2 - 3 | 299 | 89 | ||
| NM_182854.3 | NP_878274.1 | 2 - 3 | 299 | 89 | ||
| GenBank mRNA | AA278249.1 | - | 2 - 3 | 300 | 89 | |
| AK055837.1 | - | 2 - 3 | 264 | 89 | ||
| AK292650.1 | - | 2 - 3 | 258 | 89 | ||
| AK292705.1 | - | 2 - 3 | 264 | 89 | ||
| AY302441.1 | - | 2 - 3 | 201 | 89 | ||
| BC027944.1 | - | 2 - 3 | 248 | 89 | ||
| BC063423.1 | - | 2 - 3 | 240 | 89 | ||
| BC119626.1 | - | 2 - 3 | 152 | 89 | ||
More Information
Set Membership: |
|

SNX20
Genomic Map