Assay Details
| Gene Symbol: SNX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144972.1 | NP_001138444.1 | 3 - 4 | 454 | 82 | |
| GenBank mRNA | AK292650.1 | - | 3 - 4 | 413 | 82 | |
| BC063423.1 | - | 3 - 4 | 395 | 82 | ||
More Information
Set Membership: |
|

SNX20
Genomic Map