Assay Details
| Gene Symbol: GRAMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146320.1 | NP_001139792.1 | 1 - 2 | 514 | 81 | |
| NM_001146321.1 | NP_001139793.1 | 1 - 2 | 514 | 81 | ||
| NM_023927.2 | NP_076416.2 | 1 - 2 | 514 | 81 | ||
| XM_005272059.4 | XP_005272116.1 | 1 - 2 | 599 | 81 | ||
| XM_005272060.3 | XP_005272117.1 | 1 - 2 | 339 | 81 | ||
| XM_011543595.2 | XP_011541897.1 | 1 - 2 | 218 | 81 | ||
| XM_017009783.1 | XP_016865272.1 | 1 - 2 | 82 | 81 | ||
| GenBank mRNA | AK024966.1 | - | 1 - 2 | 282 | 81 | |
| AK225829.1 | - | 1 - 2 | 282 | 81 | ||
| AK293307.1 | - | 1 - 2 | 219 | 81 | ||
| AK294656.1 | - | 1 - 2 | 233 | 81 | ||
| AK296267.1 | - | 1 - 2 | 219 | 81 | ||
| AY116970.1 | - | 1 - 2 | 84 | 81 | ||
| BC008590.1 | - | 1 - 2 | 247 | 4148772 | 81 | |
| DC371647.1 | - | 1 - 2 | 514 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK293307.1, NM_001146320.1, XM_011543595.2, XM_017009783.1 transcripts.
Set Membership: |
|

GRAMD3
Genomic Map