Assay Details
| Gene Symbol: GRAMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146321.1 | NP_001139793.1 | 2 - 3 | 635 | 104 | |
| XM_005272059.4 | XP_005272116.1 | 2 - 3 | 720 | 104 | ||
| XM_005272064.3 | XP_005272121.1 | 2 - 3 | 437 | 104 | ||
| XM_011543591.2 | XP_011541893.1 | 2 - 3 | 1308 | 104 | ||
| XM_011543592.2 | XP_011541894.1 | 2 - 3 | 1304 | 104 | ||
| XM_017009781.1 | XP_016865270.1 | 2 - 3 | 195 | 104 | ||
| XM_017009782.1 | XP_016865271.1 | 2 - 3 | 437 | 104 | ||
| GenBank mRNA | AK296267.1 | - | 2 - 3 | 340 | 104 | |
More Information
Set Membership: |
|

GRAMD3
Genomic Map