Assay Details
| Gene Symbol: ZBTB33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184742.1 | NP_001171671.1 | 2 - 3 | 325 | 124 | |
| GenBank mRNA | AK312321.1 | - | 2 - 3 | 267 | 124 | |
| DB468333.1 | - | 2 - 3 | 325 | 124 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK312321.1, NM_001184742.1 transcripts.
Set Membership: |
|

ZBTB33
Genomic Map