Assay Details
| Gene Symbol: RASD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199989.1 | NP_001186918.1 | 1 - 2 | 496 | 109 | |
| GenBank mRNA | AK294073.1 | - | 1 - 2 | 496 | 109 | |
More Information
Set Membership: |
|

RASD1
Genomic Map