Assay Details
| Gene Symbol: OR5L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC136939.1 | - | 1 | 115 | 9020931 | 175 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC136939.1 transcripts.
Set Membership: |
|

OR5L1
Genomic Map