Assay Details
| Gene Symbol: HCST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007469.1 | NP_001007470.1 | 3 - 4 | 357 | 96 | |
| GenBank mRNA | AF072844.1 | - | 3 - 4 | 299 | 96 | |
| AY359058.1 | - | 3 - 4 | 298 | 96 | ||
More Information
Set Membership: |
|

HCST
Genomic Map