Assay Details
| Gene Symbol: C6orf163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010868.2 | NP_001010868.2 | 1 - 2 | 392 | 131 | |
| XM_006715370.3 | XP_006715433.1 | 1 - 2 | 2165 | 131 | ||
More Information
Set Membership: |
|

C6orf163
Genomic Map