Assay Details
| Gene Symbol: C6orf163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010868.2 | NP_001010868.2 | 2 - 3 | 492 | 173 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C6orf163| Gene Symbol: C6orf163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010868.2 | NP_001010868.2 | 2 - 3 | 492 | 173 | |
Set Membership: |
|