Assay Details
| Gene Symbol: LOC149373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256615.1 | NP_001243544.1 | 2 - 3 | 205 | 60 | |
| GenBank mRNA | BM711206.1 | - | 2 - 3 | 205 | 60 | |
More Information
Set Membership: |
|

LOC149373
Genomic Map