Assay Details
| Gene Symbol: ISCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272007.1 | NP_001258936.1 | 2 - 3 | 230 | 81 | |
| GenBank mRNA | DB501282.1 | - | 2 - 3 | 185 | 81 | |
More Information
Set Membership: |
|

ISCA2
Genomic Map