Assay Details
| Gene Symbol: PRPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002764.3 | NP_002755.1 | 2 - 3 | 511 | 58 | |
| GenBank mRNA | AK223169.1 | - | 2 - 3 | 328 | 58 | |
| AK297968.1 | - | 3 - 4 | 349 | 58 | ||
| AK312706.1 | - | 2 - 3 | 425 | 58 | ||
| BC001605.1 | - | 2 - 3 | 406 | 3542584 | 58 | |
| D00860.1 | - | 2 - 3 | 428 | 58 | ||
| DC411129.1 | - | 2 - 3 | 507 | 58 | ||
More Information
Set Membership: |
|

PRPS1
Genomic Map