Assay Details
| Gene Symbol: C6orf120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317342.1 | NP_001304271.1 | 2 | 114 | 115 | |
| GenBank mRNA | AK295962.1 | - | 2 | 89 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK295962.1, NM_001317342.1 transcripts.
Set Membership: |
|

C6orf120
Genomic Map