Assay Details
| Gene Symbol: RPL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000994.3 | NP_000985.1 | 4 | 1524 | 99 | |
| NM_001007073.1 | NP_001007074.1 | 4 | 1623 | 99 | ||
| NM_001007074.1 | NP_001007075.1 | 3 | 1643 | 99 | ||
| GenBank mRNA | CA436299.1 | - | 1 | 145 | 99 | |
More Information
Set Membership: |
|

RPL32
Genomic Map