Assay Details
| Gene Symbol: MPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000250.1 | NP_000241.1 | 8 - 9 | 1546 | 127 | |
| GenBank mRNA | BC130476.1 | - | 8 - 9 | 1420 | 40146507 | 127 |
| J02694.1 | - | 8 - 9 | 1517 | 127 | ||
| M19507.1 | - | 8 - 9 | 1546 | 127 | ||
| S56200.1 | - | 8 - 9 | 1369 | 127 | ||
| X04876.1 | - | 8 - 9 | 1532 | 127 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001727.hg.1
Set Membership: |
|

MPO
Genomic Map