Assay Details
| Gene Symbol: NYNRIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560984.2 | XP_005561041.1 | 7 - 8 | 3233 | 83 | |
| XM_015453661.1 | XP_015309147.1 | 7 - 8 | 2987 | 83 | ||
More Information
Set Membership: |
|

NYNRIN
Genomic Map