Assay Details
| Gene Symbol: GLDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559559.2 | XP_005559616.1 | 8 - 9 | 1074 | 98 | |
| XM_015452828.1 | XP_015308314.1 | 8 - 9 | 1074 | 98 | ||
More Information
Set Membership: |
|

GLDN
Genomic Map