Assay Details
| Gene Symbol: MORN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283792.1 | NP_001270721.1 | 4 - 5 | 730 | 55 | |
| XM_005572461.2 | XP_005572518.1 | 5 - 6 | 745 | 55 | ||
| GenBank mRNA | AB168617.1 | - | 4 - 5 | 730 | 55 | |
More Information
Set Membership: |
|

MORN3
Genomic Map