Assay Details
| Gene Symbol: CYP2W1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549487.1 | XP_005549544.1 | 7 - 8 | 1038 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP2W1| Gene Symbol: CYP2W1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549487.1 | XP_005549544.1 | 7 - 8 | 1038 | 82 | |
Set Membership: |
|