Assay Details
| Gene Symbol: TSPAN31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005571383.2 | XP_005571440.1 | 1 - 2 | 338 | 63 | |
| GenBank mRNA | AB173266.1 | - | 1 - 2 | 153 | 63 | |
More Information
Set Membership: |
|

TSPAN31
Genomic Map