Assay Details
| Gene Symbol: ETV5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285114.1 | NP_001272043.1 | 11 - 12 | 1339 | 70 | |
| GenBank mRNA | AB169210.1 | - | 11 - 12 | 1339 | 70 | |
More Information
Set Membership: |
|

ETV5
Genomic Map