Assay Details
Gene Symbol: PAICS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319546.1 | NP_001306475.1 | 4 - 5 | 764 | 112 | |
XM_015450435.1 | XP_015305921.1 | 6 - 7 | 815 | 112 | ||
XM_015450436.1 | XP_015305922.1 | 6 - 7 | 815 | 112 | ||
GenBank mRNA | AB173300.1 | - | 4 - 5 | 764 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
