Assay Details
Gene Symbol: HNRNPA2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283173.1 | NP_001270102.1 | 4 - 5 | 466 | 68 | |
XM_015447302.1 | XP_015302788.1 | 4 - 5 | 503 | 68 | ||
GenBank mRNA | AB173867.1 | - | 4 - 5 | 466 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
