Assay Details
| Gene Symbol: KCNJ8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570346.2 | XP_005570403.1 | 2 - 3 | 787 | 88 | |
| XM_005570347.2 | XP_005570404.1 | 2 - 3 | 682 | 88 | ||
More Information
Set Membership: |
|

KCNJ8
Genomic Map