Assay Details
| Gene Symbol: PCDH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585934.2 | XP_005585991.1 | 3 - 4 | 3052 | 105 | |
| XM_005585935.2 | XP_005585992.1 | 3 - 4 | 2760 | 105 | ||
More Information
Set Membership: |
|

PCDH8
Genomic Map