Assay Details
| Gene Symbol: ETNK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540524.2 | XP_005540581.1 | 5 - 6 | 1045 | 69 | |
| XM_005540525.2 | XP_005540582.1 | 5 - 6 | 1045 | 69 | ||
| XM_005540526.2 | XP_005540583.1 | 4 - 5 | 922 | 69 | ||
| XM_015448585.1 | XP_015304071.1 | 4 - 5 | 922 | 69 | ||
| XM_015448597.1 | XP_015304083.1 | 4 - 5 | 746 | 69 | ||
More Information
Set Membership: |
|

ETNK2
Genomic Map