Assay Details
| Gene Symbol: KLK14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590074.2 | XP_005590131.1 | 4 - 5 | 2033 | 115 | |
| XM_005590075.2 | XP_005590132.1 | 3 - 4 | 2118 | 115 | ||
| XM_005590077.2 | XP_005590134.1 | 5 - 6 | 1183 | 115 | ||
More Information
Set Membership: |
|

KLK14
Genomic Map