Assay Details
| Gene Symbol: TMEM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283269.1 | NP_001270198.1 | 1 - 2 | 400 | 64 | |
| XM_005543294.2 | XP_005543351.1 | 1 - 2 | 459 | 64 | ||
| GenBank mRNA | AB168526.1 | - | 1 - 2 | 400 | 64 | |
| AB170371.1 | - | 1 - 2 | 371 | 64 | ||
More Information
Set Membership: |
|

TMEM59
Genomic Map