Assay Details
Gene Symbol: MED26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015440527.1 | XP_015296013.1 | 2 - 3 | 202 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
